Anti-Viperin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-03971
102234-718EA 417.23 USD
102234-718
Anti-Viperin Rabbit Polyclonal Antibody
Antibodies
The Viperin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Viperin. This antibody reacts with human, mouse, rat, porcine, bovine, canine, chicken, chinese hamster, equine, primate, rabbit. The Viperin Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: Viperin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Chinese Hamster, Equine, Primate, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR