Anti-VIP Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-65658
102114-912EA 299 USD
102114-912
Anti-VIP Rabbit Polyclonal Antibody
Antibodies
The VIP Antibody from Novus Biologicals is a rabbit polyclonal antibody to VIP. This antibody reacts with human. The VIP Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: VIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR