Anti-VEGF Rabbit Polyclonal Antibody (DyLight 550)

Supplier: Novus Biologicals
NB100-2381R
102107-260EA 626.46 USD
102107-260
Anti-VEGF Rabbit Polyclonal Antibody (DyLight 550)
Antibodies
The VEGF Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to VEGF. This antibody reacts with human, mouse, rat, bovine, canine, chicken, equine, guinea pig. The VEGF Antibody [DyLight 550] has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: VEGF
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR