Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VEGF Rabbit Polyclonal Antibody (Biotin)

Supplier: Novus Biologicals
NB100-2381B
102107-250
Anti-VEGF Rabbit Polyclonal Antibody (Biotin)
The VEGF Antibody [Biotin] from Novus Biologicals is a rabbit polyclonal antibody to VEGF. This antibody reacts with human, mouse, rat, bovine, canine, chicken, equine, guinea pig. The VEGF Antibody [Biotin] has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: VEGF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Guinea Pig

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty