Anti-VEGF R2/KDR/Flk-1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Novus Biologicals
NB100-529H
102109-512EA 614.15 USD
102109-512
Anti-VEGF R2/KDR/Flk-1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
The VEGF R2 / KDR / Flk-1 Antibody [HRP] from Novus Biologicals is a rabbit polyclonal antibody to VEGF R2 / KDR / Flk-1. This antibody reacts with human, mouse. The VEGF R2 / KDR / Flk-1 Antibody [HRP] has been validated for the following applications: Western Blot.

Type: Primary
Antigen: VEGF R2/KDR/Flk-1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR