Anti-UROS Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-32485
102165-986EA 635.51 USD
102165-986
Anti-UROS Rabbit Polyclonal Antibody
Antibodies
The UROS Antibody from Novus Biologicals is a rabbit polyclonal antibody to UROS. This antibody reacts with human. The UROS Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: UROS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR