Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Uromodulin Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-58867
102191-458
Anti-Uromodulin Rabbit Polyclonal Antibody
The Uromodulin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Uromodulin. This antibody reacts with human. The Uromodulin Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Uromodulin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty