Anti-UNG Rabbit Polyclonal Antibody

NBP1-77259
102211-212EA 466.15 USD
102211-212
Anti-UNG Rabbit Polyclonal Antibody
The UNG Antibody from Novus Biologicals is a rabbit polyclonal antibody to UNG. This antibody reacts with human, mouse, rat. The UNG Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Recommended Dilutions: Immunohistochemistry (Parafin): 2 ?g/ml

Type: Primary
Antigen: UNG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR