Anti-UQCR10 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-62349
102195-522EA 564.74 USD
102195-522
Anti-UQCR10 Rabbit Polyclonal Antibody
Antibodies
The UQCR10 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UQCR10. This antibody reacts with human. The UQCR10 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: UQCR10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR