Anti-UGT2B4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-69604
102199-160EA 564.74 USD
102199-160
Anti-UGT2B4 Rabbit Polyclonal Antibody
Antibodies
The UGT2B4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UGT2B4. This antibody reacts with human. The UGT2B4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry.

Type: Primary
Antigen: UGT2B4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR