Anti-UGT1A6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-69707
102199-330EA 564.74 USD
102199-330
Anti-UGT1A6 Rabbit Polyclonal Antibody
Antibodies
The UGT1A6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UGT1A6. This antibody reacts with human. The UGT1A6 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry.

Type: Primary
Antigen: UGT1A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR