Anti-UFD1L Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-03338
102155-844EA 635.51 USD
102155-844
Anti-UFD1L Rabbit Polyclonal Antibody
Antibodies
The UFD1L Antibody from Novus Biologicals is a rabbit polyclonal antibody to UFD1L. This antibody reacts with human. The UFD1L Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: UFD1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR