Anti-UCP4 Rabbit Polyclonal Antibody

NB100-66495
102115-358EA 423.08 USD
102115-358
Anti-UCP4 Rabbit Polyclonal Antibody
The UCP4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UCP4. This antibody reacts with non-species specific. The UCP4 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: UCP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Non-species specific
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR