Anti-UCMA Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-72115
102201-936EA 635.51 USD
102201-936
Anti-UCMA Rabbit Polyclonal Antibody
Antibodies
The UCMA Antibody from Novus Biologicals is a rabbit polyclonal antibody to UCMA. This antibody reacts with human, mouse. The UCMA Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 5 ?g/ml

Type: Primary
Antigen: UCMA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR