Anti-UCH-L1/PGP9.5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-1616
102105-648EA 257 USD
102105-648
Anti-UCH-L1/PGP9.5 Rabbit Polyclonal Antibody
Antibodies
The UCH-L1 / PGP9.5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UCH-L1 / PGP9.5. This antibody reacts with human, mouse, rat, porcine. The UCH-L1 / PGP9.5 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: UCH-L1/PGP9.5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Porcine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR