Anti-UCH-L1/PGP9.5 Rabbit Polyclonal Antibody (DyLight 488)

Supplier: Novus Biologicals
NB300-675G
102130-960EA 614.15 USD
102130-960
Anti-UCH-L1/PGP9.5 Rabbit Polyclonal Antibody (DyLight 488)
Antibodies
The UCH-L1 / PGP9.5 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to UCH-L1 / PGP9.5. This antibody reacts with human, mouse. The UCH-L1 / PGP9.5 Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: UCH-L1/PGP9.5
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR