Anti-UBXN6 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-98481
102223-278EA 564.74 USD
102223-278
Anti-UBXN6 Rabbit Polyclonal Antibody
Antibodies
The UBXN6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UBXN6. This antibody reacts with mouse. The UBXN6 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: UBXN6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR