Anti-UBN1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-49983
102179-858EA 635.51 USD
102179-858
Anti-UBN1 Rabbit Polyclonal Antibody
Antibodies
The UBN1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UBN1. This antibody reacts with human. The UBN1 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation (Negative).

Type: Primary
Antigen: UBN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR