Anti-Ubiquitin Rabbit Polyclonal Antibody

NBP1-23015
102160-844EA 564.62 USD
102160-844
Anti-Ubiquitin Rabbit Polyclonal Antibody
The Ubiquitin Antibody from Novus Biologicals is a rabbit polyclonal antibody to Ubiquitin. This antibody reacts with human, mouse. The Ubiquitin Antibody has been validated for the following applications: Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100 - 1:500

Type: Primary
Antigen: Ubiquitin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR