Anti-E6AP/UBE3A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB500-239
102132-092EA 635.51 USD
102132-092
Anti-E6AP/UBE3A Rabbit Polyclonal Antibody
Antibodies
The E6AP / UBE3A Antibody from Novus Biologicals is a rabbit polyclonal antibody to E6AP / UBE3A. This antibody reacts with human, mouse. The E6AP / UBE3A Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: E6AP/UBE3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR