Anti-Uev1a/UBE2V1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-56397
102109-954EA 279 USD
102109-954
Anti-Uev1a/UBE2V1 Rabbit Polyclonal Antibody
Antibodies
The Uev1a / UBE2V1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Uev1a / UBE2V1. This antibody reacts with human. The Uev1a / UBE2V1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Uev1a/UBE2V1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR