Anti-UBE2Q2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-49694
102179-438EA 635.51 USD
102179-438
Anti-UBE2Q2 Rabbit Polyclonal Antibody
Antibodies
The UBE2Q2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UBE2Q2. This antibody reacts with human, mouse. The UBE2Q2 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: UBE2Q2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR