Anti-UBE2N/Ubc13 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-76593
102208-632EA 564.74 USD
102208-632
Anti-UBE2N/Ubc13 Rabbit Polyclonal Antibody
Antibodies
The UBE2N / Ubc13 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UBE2N / Ubc13. This antibody reacts with human, mouse, rat. The UBE2N / Ubc13 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 2 ?g/ml

Type: Primary
Antigen: UBE2N/Ubc13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR