Anti-UBE2N/Ubc13 Rabbit Polyclonal Antibody

NBP1-54894
102184-746EA 423.08 USD
102184-746
Anti-UBE2N/Ubc13 Rabbit Polyclonal Antibody
The UBE2N / Ubc13 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UBE2N / Ubc13. This antibody reacts with human, mouse, non-species specific. The UBE2N / Ubc13 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: UBE2N/Ubc13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Non-species specific
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR