Anti-UBE2J1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59757
102193-200EA 564.74 USD
102193-200
Anti-UBE2J1 Rabbit Polyclonal Antibody
Antibodies
The UBE2J1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UBE2J1. This antibody reacts with human. The UBE2J1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: UBE2J1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR