Anti-UBE2G2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-32729
102166-306EA 635.51 USD
102166-306
Anti-UBE2G2 Rabbit Polyclonal Antibody
Antibodies
The UBE2G2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to UBE2G2. This antibody reacts with human. The UBE2G2 Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: UBE2G2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR