Anti-UbcH10/UBE2C Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-58165
102191-018EA 564.74 USD
102191-018
Anti-UbcH10/UBE2C Rabbit Polyclonal Antibody
Antibodies
The UbcH10 / UBE2C Antibody from Novus Biologicals is a rabbit polyclonal antibody to UbcH10 / UBE2C. This antibody reacts with human. The UbcH10 / UBE2C Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: UbcH10/UBE2C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR