Anti-UBE2A Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB100-554
102109-750EA 635.51 USD
102109-750
Anti-UBE2A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal UBE2A Antibody. Tested Applications: Western Blot. Tested Reactivity: Human.

Type: Primary
Antigen: UBE2A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR