Anti-U2AF35 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-32515
102166-026EA 635.51 USD
102166-026
Anti-U2AF35 Rabbit Polyclonal Antibody
Antibodies
The U2AF35 Antibody from Novus Biologicals is a rabbit polyclonal antibody to U2AF35. This antibody reacts with human, mouse. The U2AF35 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:1000

Type: Primary
Antigen: U2AF35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR