Anti-Dtk/TYRO3 Rabbit Polyclonal Antibody

NB100-2311
102107-074EA 536.92 USD
102107-074
Anti-Dtk/TYRO3 Rabbit Polyclonal Antibody
The Dtk / TYRO3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Dtk / TYRO3. This antibody reacts with human. The Dtk / TYRO3 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: Dtk/TYRO3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR