Anti-TUG Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-28710
102162-568EA 635.51 USD
102162-568
Anti-TUG Rabbit Polyclonal Antibody
Antibodies
The TUG Antibody from Novus Biologicals is a rabbit polyclonal antibody to TUG. This antibody reacts with human. The TUG Antibody has been validated for the following applications: Immunoprecipitation, Western Blot (Negative).

Type: Primary
Antigen: TUG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR