Anti-TTK Rabbit Polyclonal Antibody

NB100-462
102109-234EA 536.92 USD
102109-234
Anti-TTK Rabbit Polyclonal Antibody
The TTK Antibody from Novus Biologicals is a rabbit polyclonal antibody to TTK. This antibody reacts with human, mouse. The TTK Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: TTK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR