Anti-TTI1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-79034
102213-746EA 635.51 USD
102213-746
Anti-TTI1 Rabbit Polyclonal Antibody
Antibodies
The TTI1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TTI1. This antibody reacts with human. The TTI1 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation, Proximity Ligation Assay.

Type: Primary
Antigen: TTI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR