Anti-TTF2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-32842
102166-458EA 635.51 USD
102166-458
Anti-TTF2 Rabbit Polyclonal Antibody
Antibodies
The TTF2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TTF2. This antibody reacts with human. The TTF2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TTF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR