Anti-TTC9C Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-55306
102185-556EA 564.74 USD
102185-556
Anti-TTC9C Rabbit Polyclonal Antibody
Antibodies
The TTC9C Antibody from Novus Biologicals is a rabbit polyclonal antibody to TTC9C. This antibody reacts with human. The TTC9C Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TTC9C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR