Anti-TSKS Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-54878
102184-716EA 564.74 USD
102184-716
Anti-TSKS Rabbit Polyclonal Antibody
Antibodies
The TSKS Antibody from Novus Biologicals is a rabbit polyclonal antibody to TSKS. This antibody reacts with human. The TSKS Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TSKS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR