Anti-TSKS Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB200-147
102129-138EA 279 USD
102129-138
Anti-TSKS Rabbit Polyclonal Antibody
Antibodies
The TSKS Antibody from Novus Biologicals is a rabbit polyclonal antibody to TSKS. This antibody reacts with human. The TSKS Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: TSKS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR