Anti-TSH R Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-53167
102183-790EA 578.89 USD
102183-790
Anti-TSH R Rabbit Polyclonal Antibody
Antibodies
The TSH R Antibody from Novus Biologicals is a rabbit polyclonal antibody to TSH R. This antibody reacts with human. The TSH R Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TSH R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR