Anti-TSC2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-76619
102208-728EA 564.74 USD
102208-728
Anti-TSC2 Rabbit Polyclonal Antibody
Antibodies
The TSC2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TSC2. This antibody reacts with human, mouse. The TSC2 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TSC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR