Anti-Tryptophan hydroxylase 2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-77556 NBP1-77556SS
102211-792EA 208 USD
102211-792 102211-790
Anti-Tryptophan hydroxylase 2 Rabbit Polyclonal Antibody
Antibodies
The Tryptophan hydroxylase 2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Tryptophan hydroxylase 2. This antibody reacts with human. The Tryptophan hydroxylase 2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Tryptophan hydroxylase 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR