Anti-Tryptophan Hydroxylase 1/TPH-1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NB300-260
102130-406EA 491.08 USD
102130-406
Anti-Tryptophan Hydroxylase 1/TPH-1 Rabbit Polyclonal Antibody
Antibodies
The Tryptophan Hydroxylase 1 / TPH-1 [p Ser58] Antibody from Novus Biologicals is a rabbit polyclonal antibody to Tryptophan Hydroxylase 1 / TPH-1. This antibody reacts with rabbit. The Tryptophan Hydroxylase 1 / TPH-1 [p Ser58] Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Tryptophan Hydroxylase 1/TPH-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR