Anti-TRPM7 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-46829
102176-542EA 635.51 USD
102176-542
Anti-TRPM7 Rabbit Polyclonal Antibody
Antibodies
The TRPM7 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRPM7. This antibody reacts with human. The TRPM7 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: TRPM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR