Anti-TRPM3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-80210
102216-068EA 564.74 USD
102216-068
Anti-TRPM3 Rabbit Polyclonal Antibody
Antibodies
The TRPM3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TRPM3. This antibody reacts with human. The TRPM3 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TRPM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR