Anti-TRPM2 Rabbit Polyclonal Antibody (DyLight 488)

Supplier: Novus Biologicals
NB110-81601G
102125-558EA 614.15 USD
102125-558
Anti-TRPM2 Rabbit Polyclonal Antibody (DyLight 488)
Antibodies
The TRPM2 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to TRPM2. This antibody reacts with mouse, rat. The TRPM2 Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Frozen.

Type: Primary
Antigen: TRPM2
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR