Anti-TRPA1 Rabbit Polyclonal Antibody (DyLight 550)

Supplier: Novus Biologicals
NB110-40763R
102122-068EA 700.31 USD
102122-068
Anti-TRPA1 Rabbit Polyclonal Antibody (DyLight 550)
Antibodies
The TRPA1 Antibody [DyLight 550] from Novus Biologicals is a rabbit polyclonal antibody to TRPA1. This antibody reacts with human, mouse, guinea pig. The TRPA1 Antibody [DyLight 550] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: TRPA1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR