Anti-TRPA1 Rabbit Polyclonal Antibody (DyLight 488)

Supplier: Novus Biologicals
NB110-40763G
102122-064EA 700.31 USD
102122-064
Anti-TRPA1 Rabbit Polyclonal Antibody (DyLight 488)
Antibodies
The TRPA1 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to TRPA1. This antibody reacts with human, mouse, guinea pig. The TRPA1 Antibody [DyLight 488] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:250

Type: Primary
Antigen: TRPA1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR