Anti-Troponin I Type 1 (slow skeletal) Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-56641
102188-206EA 635.51 USD
102188-206
Anti-Troponin I Type 1 (slow skeletal) Rabbit Polyclonal Antibody
Antibodies
The Troponin I Type 1 (slow skeletal) Antibody from Novus Biologicals is a rabbit polyclonal antibody to Troponin I Type 1 (slow skeletal). This antibody reacts with human. The Troponin I Type 1 (slow skeletal) Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 4-8 ?g/ml

Type: Primary
Antigen: Troponin I Type 1 (slow skeletal)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR