Anti-TMEM231 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-59823
102193-330EA 564.74 USD
102193-330
Anti-TMEM231 Rabbit Polyclonal Antibody
Antibodies
The TMEM231 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TMEM231. This antibody reacts with human. The TMEM231 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: TMEM231
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR