Anti-TMEM184B Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-79196
102214-040EA 564.74 USD
102214-040
Anti-TMEM184B Rabbit Polyclonal Antibody
Antibodies
The TMEM184B Antibody from Novus Biologicals is a rabbit polyclonal antibody to TMEM184B. This antibody reacts with rat. The TMEM184B Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: TMEM184B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR