Anti-TMEM163 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-06608
102157-326EA 564.74 USD
102157-326
Anti-TMEM163 Rabbit Polyclonal Antibody
Antibodies
The TMEM163 Antibody from Novus Biologicals is a rabbit polyclonal antibody to TMEM163. This antibody reacts with human. The TMEM163 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:200

Type: Primary
Antigen: TMEM163
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR